Imprimer

Anti-ATG12 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 43CT73.3.5.5.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030788-PEEA 720 EUR
USBI030788-PE
Anti-ATG12 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 43CT73.3.5.5.4]
Anticorps
Recommended dilution: FLISA: 1:10000; Western Blot: 1:200-2000; Immunochemistry: 1:50-200; Immunofluorescence:1:10-50
Do not freeze! Light sensitive

Type: Primary
Antigen: ATG12 (autophagy related 12)
Clonality: Monoclonal
Clone: 43CT73.3.5.5.4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR