Imprimer

Anti-ATG7 Mouse Monoclonal Antibody [clone: 29CT10.3.12]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030796EA 535 EUR
USBI030796
Anti-ATG7 Mouse Monoclonal Antibody [clone: 29CT10.3.12]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-2000

Type: Primary
Antigen: ATG7 (autophagy related 7)
Clonality: Monoclonal
Clone: 29CT10.3.12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR