Imprimer

Anti-BAX Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 557CT5.2.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030802-PEEA 711 EUR
USBI030802-PE
Anti-BAX Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 557CT5.2.1]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100
Do not freeze! Light sensitive

Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: Monoclonal
Clone: 557CT5.2.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR