Imprimer

Anti-BCL6 Mouse Monoclonal Antibody (Biotin) [clone: 305CT16.3.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030804-BIOTINEA 711 EUR
USBI030804-BIOTIN
Anti-BCL6 Mouse Monoclonal Antibody (Biotin) [clone: 305CT16.3.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: BCL6 (B-cell CLL/lymphoma 6)
Clonality: Monoclonal
Clone: 305CT16.3.3
Conjugation: Biotin
Epitope: C-terminal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR