Imprimer

Anti-BECN1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 15CT26]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030805-FITCEA 678 EUR
USBI030805-FITC
Anti-BECN1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 15CT26]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: Monoclonal
Clone: 15CT26
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR