Imprimer

Anti-BECN1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 15CT26]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030805-PEEA 619 EUR
USBI030805-PE
Anti-BECN1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 15CT26]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50; Immunochemistry: 1:50-1:200
Do not freeze! Light sensitive

Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: Monoclonal
Clone: 15CT26
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR