Imprimer

Anti-REL Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 318CT41.3.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030818-FITCEA 852 EUR
USBI030818-FITC
Anti-REL Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 318CT41.3.2]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:120-1000

Type: Primary
Antigen: REL (v-rel avian reticuloendotheliosis viral oncogene homolog)
Clonality: Monoclonal
Clone: 318CT41.3.2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR