Imprimer

Anti-CALCA Mouse Monoclonal Antibody [clone: 406CT21.4.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030825EA 527 EUR
USBI030825
Anti-CALCA Mouse Monoclonal Antibody [clone: 406CT21.4.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-8000

Type: Primary
Antigen: CALCA (calcitonin-related polypeptide alpha)
Clonality: Monoclonal
Clone: 406CT21.4.3
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR