Imprimer

Anti-SDC1 Mouse Monoclonal Antibody [clone: 480CT5.4.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030839EA 420 EUR
USBI030839
Anti-SDC1 Mouse Monoclonal Antibody [clone: 480CT5.4.3]
Anticorps
Recommended dilution: Western Blot: 1:100-1:500; Immunofluorescence: 1:10-1:50; Flow Cytometry: 1:10-150

Type: Primary
Antigen: SDC1 (syndecan 1)
Clonality: Monoclonal
Clone: 480CT5.4.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR