Imprimer

Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7G7B6]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030844-PEEA 562 EUR
USBI030844-PE
Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7G7B6]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:250
Do not freeze! Light sensitive

Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: Monoclonal
Clone: 7G7B6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR