Imprimer

Anti-CD34 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 132CT29.1.5]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030848-HRPEA 852 EUR
USBI030848-HRP
Anti-CD34 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 132CT29.1.5]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: Monoclonal
Clone: 132CT29.1.5
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR