Imprimer

Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030849EA 527 EUR
USBI030849
Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4]
Anticorps
  • Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-800

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: Monoclonal
Clone: OKT4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR