Imprimer

Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Hermes-3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030854-FITCEA 852 EUR
USBI030854-FITC
Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Hermes-3]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: Hermes-3
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR