Imprimer

Anti-CD55 Mouse Monoclonal Antibody [clone: 561CT4.2.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030855EA 527 EUR
USBI030855
Anti-CD55 Mouse Monoclonal Antibody [clone: 561CT4.2.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-8000

Type: Primary
Antigen: CD55 (CD55 molecule, decay accelerating factor for complement (Cromer blood group))
Clonality: Monoclonal
Clone: 561CT4.2.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR