Imprimer

Anti-CD68 Mouse Monoclonal Antibody [clone: 404CT9.2.5]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030859EA 527 EUR
USBI030859
Anti-CD68 Mouse Monoclonal Antibody [clone: 404CT9.2.5]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-4000

Type: Primary
Antigen: CD68 (CD68 molecule)
Clonality: Monoclonal
Clone: 404CT9.2.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR