Imprimer

Anti-CD79A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 299CT22.1.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030862-APCEA 711 EUR
USBI030862-APC
Anti-CD79A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 299CT22.1.2]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: CD79A (CD79a molecule, immunoglobulin-associated alpha)
Clonality: Monoclonal
Clone: 299CT22.1.2
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR