Imprimer

Anti-CDCA8 Mouse Monoclonal Antibody [clone: 181CT41.2.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030868EA 495 EUR
USBI030868
Anti-CDCA8 Mouse Monoclonal Antibody [clone: 181CT41.2.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CDCA8 (cell division cycle associated 8)
Clonality: Monoclonal
Clone: 181CT41.2.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR