Imprimer

Anti-CHEK1 Mouse Monoclonal Antibody (Biotin) [clone: 2G1D5]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030874-BIOTINEA 619 EUR
USBI030874-BIOTIN
Anti-CHEK1 Mouse Monoclonal Antibody (Biotin) [clone: 2G1D5]
Anticorps
Recommended dilution: ELISA: 1:10000; Western Blot: 1:1000-4000

Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: Monoclonal
Clone: 2G1D5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR