Imprimer

Anti-CLIC5 Mouse Monoclonal Antibody (Biotin) [clone: 16CT34.4.8]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030877-BIOTINEA 711 EUR
USBI030877-BIOTIN
Anti-CLIC5 Mouse Monoclonal Antibody (Biotin) [clone: 16CT34.4.8]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: CLIC5 (chloride intracellular channel 5)
Clonality: Monoclonal
Clone: 16CT34.4.8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR