Imprimer

Anti-COL3A1 Mouse Monoclonal Antibody [clone: 539CT14.1.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030884EA 495 EUR
USBI030884
Anti-COL3A1 Mouse Monoclonal Antibody [clone: 539CT14.1.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: COL3A1 (collagen, type III, alpha 1)
Clonality: Monoclonal
Clone: 539CT14.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR