Imprimer

Anti-COMT Mouse Monoclonal Antibody [clone: 630CT1.7.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030887EA 527 EUR
USBI030887
Anti-COMT Mouse Monoclonal Antibody [clone: 630CT1.7.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-400

Type: Primary
Antigen: COMT (catechol-O-methyltransferase)
Clonality: Monoclonal
Clone: 630CT1.7.1
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR