Imprimer

Anti-CTGF Mouse Monoclonal Antibody (Biotin) [clone: 558CT34.4.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030895-BIOTINEA 711 EUR
USBI030895-BIOTIN
Anti-CTGF Mouse Monoclonal Antibody (Biotin) [clone: 558CT34.4.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: Monoclonal
Clone: 558CT34.4.4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR