Imprimer

Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 516CT4.3.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030896EA 527 EUR
USBI030896
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 516CT4.3.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:200-1600

Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: Monoclonal
Clone: 516CT4.3.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR