Imprimer

Anti-CYK18 Mouse Monoclonal Antibody [clone: 695CT17.2.6]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030901EA 587 EUR
USBI030901
Anti-CYK18 Mouse Monoclonal Antibody [clone: 695CT17.2.6]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-8000

Type: Primary
Antigen: CYK18
Clonality: Monoclonal
Clone: 695CT17.2.6
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG3
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR