Imprimer

Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 499CT13.2.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030905EA 587 EUR
USBI030905
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 499CT13.2.2]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: CYP1A2 (cytochrome P450, family 1, subfamily A, polypeptide 2)
Clonality: Monoclonal
Clone: 499CT13.2.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR