Imprimer

Anti-DPPA2 Mouse Monoclonal Antibody [clone: 210CT5.14.6.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030920EA 527 EUR
USBI030920
Anti-DPPA2 Mouse Monoclonal Antibody [clone: 210CT5.14.6.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-16000

Type: Primary
Antigen: DPPA2 (developmental pluripotency associated 2)
Clonality: Monoclonal
Clone: 210CT5.14.6.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR