Imprimer

Anti-EGFR Mouse Monoclonal Antibody (Biotin) [clone: 688CT33.1.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030930-BIOTINEA 720 EUR
USBI030930-BIOTIN
Anti-EGFR Mouse Monoclonal Antibody (Biotin) [clone: 688CT33.1.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000

Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: Monoclonal
Clone: 688CT33.1.3
Conjugation: Biotin
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR