Imprimer

Anti-EIF4E Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 163CT48.1.9]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030936-FITCEA 720 EUR
USBI030936-FITC
Anti-EIF4E Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 163CT48.1.9]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50

Type: Primary
Antigen: EIF4E (eukaryotic translation initiation factor 4E)
Clonality: Monoclonal
Clone: 163CT48.1.9
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR