Imprimer

Anti-EIF4E2 Mouse Monoclonal Antibody [clone: 225CT3.1.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030937EA 527 EUR
USBI030937
Anti-EIF4E2 Mouse Monoclonal Antibody [clone: 225CT3.1.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1600

Type: Primary
Antigen: EIF4E2 (eukaryotic translation initiation factor 4E family member 2)
Clonality: Monoclonal
Clone: 225CT3.1.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR