Imprimer

Anti-ELAVL1 Mouse Monoclonal Antibody (Biotin) [clone: 224CT6.5.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030942-BIOTINEA 711 EUR
USBI030942-BIOTIN
Anti-ELAVL1 Mouse Monoclonal Antibody (Biotin) [clone: 224CT6.5.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ELAVL1 (ELAV like RNA binding protein 1)
Clonality: Monoclonal
Clone: 224CT6.5.3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR