Imprimer

Anti-ESR2 Mouse Monoclonal Antibody [clone: 555CT18.5.5]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030953EA 587 EUR
USBI030953
Anti-ESR2 Mouse Monoclonal Antibody [clone: 555CT18.5.5]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-4000

Type: Primary
Antigen: ESR2 (estrogen receptor 2 (ER beta))
Clonality: Monoclonal
Clone: 555CT18.5.5
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR