Imprimer

Anti-FAU Mouse Monoclonal Antibody [clone: 291CT8.1.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030957EA 587 EUR
USBI030957
Anti-FAU Mouse Monoclonal Antibody [clone: 291CT8.1.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-8000

Type: Primary
Antigen: FAU (Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed)
Clonality: Monoclonal
Clone: 291CT8.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR