Imprimer

Anti-FGA Mouse Monoclonal Antibody [clone: 471CT12.1.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030961EA 527 EUR
USBI030961
Anti-FGA Mouse Monoclonal Antibody [clone: 471CT12.1.2]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: FGA (fibrinogen alpha chain)
Clonality: Monoclonal
Clone: 471CT12.1.2
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR