Imprimer

Anti-FKRP Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 386CT13.1.9]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030965-FITCEA 711 EUR
USBI030965-FITC
Anti-FKRP Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 386CT13.1.9]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: FKRP (fukutin related protein)
Clonality: Monoclonal
Clone: 386CT13.1.9
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR