Imprimer

Anti-FOXP2 Mouse Monoclonal Antibody [clone: 533CT26.1.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030970EA 527 EUR
USBI030970
Anti-FOXP2 Mouse Monoclonal Antibody [clone: 533CT26.1.2]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1600

Type: Primary
Antigen: FOXP2 (forkhead box P2)
Clonality: Monoclonal
Clone: 533CT26.1.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR