Imprimer

Anti-GAB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1AT1979]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030976-APCEA 852 EUR
USBI030976-APC
Anti-GAB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1AT1979]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:200-1000; Immunoprecipitation: 2-5 ?g

Type: Primary
Antigen: GAB1 (GRB2-associated binding protein 1)
Clonality: Monoclonal
Clone: 1AT1979
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR