Imprimer

Anti-GATA4 Mouse Monoclonal Antibody [clone: 570CT10.5.7]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030983EA 587 EUR
USBI030983
Anti-GATA4 Mouse Monoclonal Antibody [clone: 570CT10.5.7]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-400

Type: Primary
Antigen: GATA4 (GATA binding protein 4)
Clonality: Monoclonal
Clone: 570CT10.5.7
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR