Imprimer

Anti-MLST8 Mouse Monoclonal Antibody [clone: 211CT7.1.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030984EA 527 EUR
USBI030984
Anti-MLST8 Mouse Monoclonal Antibody [clone: 211CT7.1.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-8000

Type: Primary
Antigen: MLST8 (MTOR associated protein, LST8 homolog (S. cerevisiae))
Clonality: Monoclonal
Clone: 211CT7.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR