Imprimer

Anti-GFAP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 183CT3.1.5]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030990-APCEA 720 EUR
USBI030990-APC
Anti-GFAP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 183CT3.1.5]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: Monoclonal
Clone: 183CT3.1.5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR