Imprimer

Anti-GFP Mouse Monoclonal Antibody [clone: 168AT1211]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030991EA 544 EUR
USBI030991
Anti-GFP Mouse Monoclonal Antibody [clone: 168AT1211]
Anticorps
Recommended dilution: ELISA: 1:3000; Western Blot: 1:2000; Immunochemistry: 1:50-100; Immunofluorescence: 1:50-100

Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: Monoclonal
Clone: 168AT1211
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR