Imprimer

Anti-GJC2 Mouse Monoclonal Antibody [clone: 391CT6.4.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030993EA 527 EUR
USBI030993
Anti-GJC2 Mouse Monoclonal Antibody [clone: 391CT6.4.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-8000

Type: Primary
Antigen: GJC2 (gap junction protein, gamma 2, 47kDa)
Clonality: Monoclonal
Clone: 391CT6.4.3
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR