Imprimer

Anti-GCG Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 329CT36.10.10]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030996-APCEA 711 EUR
USBI030996-APC
Anti-GCG Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 329CT36.10.10]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: GCG (glucagon)
Clonality: Monoclonal
Clone: 329CT36.10.10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR