Imprimer

Anti-Glutathione S Transferase Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 21CT54.13.1 / 21CT54.13.7]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031004-APEA 720 EUR
USBI031004-AP
Anti-Glutathione S Transferase Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 21CT54.13.1 / 21CT54.13.7]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Tag
Antigen: Glutathione S Transferase
Clonality: Monoclonal
Clone: 21CT54.13.1 / 21CT54.13.7
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR