Imprimer

Anti-Glutathione S Transferase Mouse Monoclonal Antibody (Biotin) [clone: 9AT106]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031005-BIOTINEA 720 EUR
USBI031005-BIOTIN
Anti-Glutathione S Transferase Mouse Monoclonal Antibody (Biotin) [clone: 9AT106]
Anticorps
Recommended dilution: ELISA: 1:3000; Western Blot: 1:2000

Type: Tag
Antigen: Glutathione S Transferase
Clonality: Monoclonal
Clone: 9AT106
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR