Imprimer

Anti-HGF Mouse Monoclonal Antibody [clone: 489CT6.12.6]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031014EA 495 EUR
USBI031014
Anti-HGF Mouse Monoclonal Antibody [clone: 489CT6.12.6]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: HGF (hepatocyte growth factor (hepapoietin A; scatter factor))
Clonality: Monoclonal
Clone: 489CT6.12.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR