Imprimer

Anti-HLA-A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7G7F9]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031022-PEEA 711 EUR
USBI031022-PE
Anti-HLA-A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7G7F9]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250
Do not freeze!

Type: Primary
Antigen: HLA-A (major histocompatibility complex, class I, A)
Clonality: Monoclonal
Clone: 7G7F9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR