Imprimer

Anti-HLA-B Mouse Monoclonal Antibody [clone: 528CT10.4.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031023EA 527 EUR
USBI031023
Anti-HLA-B Mouse Monoclonal Antibody [clone: 528CT10.4.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: HLA-B (major histocompatibility complex, class I, B)
Clonality: Monoclonal
Clone: 528CT10.4.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR