Imprimer

Anti-HNRNPA2B1 Mouse Monoclonal Antibody (Biotin) [clone: 277CT1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031032-BIOTINEA 711 EUR
USBI031032-BIOTIN
Anti-HNRNPA2B1 Mouse Monoclonal Antibody (Biotin) [clone: 277CT1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:30

Type: Primary
Antigen: HNRNPA2B1 (heterogeneous nuclear ribonucleoprotein A2/B1)
Clonality: Monoclonal
Clone: 277CT1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR