Imprimer

Anti-HSPA1A Mouse Monoclonal Antibody (Biotin) [clone: 201CT8.2.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031037-BIOTINEA 711 EUR
USBI031037-BIOTIN
Anti-HSPA1A Mouse Monoclonal Antibody (Biotin) [clone: 201CT8.2.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-2000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: HSPA1A (heat shock 70kDa protein 1A)
Clonality: Monoclonal
Clone: 201CT8.2.3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR