Imprimer

Anti-IKBKB Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 62AT216]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031045-HRPEA 720 EUR
USBI031045-HRP
Anti-IKBKB Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 62AT216]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: Monoclonal
Clone: 62AT216
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR